missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Modified Letheen Broth
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Oxoid Letheen Broth, Modified is used for the evaluation of germicide performance for sanitation of food utensils and to evaluate cosmetics.
Specifications
Specifications
| Description | Letheen Broth, Modified |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
For Laboratory Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction