missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Modified Czapek Dox Agar (Dehydrated)

Product Code. 15394772
Click to view available options
Quantity:
500 g
Unit Size:
45.4g
This item is not returnable. View return policy

Product Code. 15394772

Brand: Thermo Scientific™ CM0097B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate fungi and bacteria which utilize sodium nitrate as the sole source of nitrogen with this medium.

Cultivate fungi and bacteria which utilize sodium nitrate as the sole source of nitrogen with Thermo Scientific™ Oxoid™ Modified Czapek Dox Agar (Dehydrated). This solid defined medium is one of the most useful media for the general cultivation of fungi. In the Modified Czapek Dox Agar magnesium glycerophosphate and potassium sulphate replace the magnesium sulphate and potassium phosphate of the original. This modification prevents the precipitation of magnesium phosphate. The medium is also a highly satisfactory substrate for chlamydospore production by Candida albicans 1.

Use Modified Czapek Dox Agar for cultivation of fungi and bacteria which utilize sodium nitrate as the sole source of nitrogen.

Modified Czapek Dox Agar has shown improved chlamydospore production as compared to the original formulation1.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Czapek Dox Agar Modified
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.0L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.