missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories

Product Code. 17108234
Change view
Click to view available options
Description:
Microbact Holding Tray 12A/12B
Microbact Holding Tray 24E/12E
Microbact Mineral Oil
Microbact Oxidase Strip
Microbact Reagent Indole-Kovacs
Microbact Reagent Nitrate A
Microbact Reagent Nitrate B
Microbact Reagent Set D
Microbact Reagent Spot Indole DMACA
Microbact Reagent TDA
Microbact Reagent VP I
Microbact Reagent VP II
Quantity:
1 Bottle
1 Tray
10 mL/Bottle
100/Pk.
50/Pk.
6 x 10 mL Bottles
Unit Size:
10mL
50 tests
50mL
Each
Pack of 10
Pack of 100
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Unit Size
17108234 Microbact Oxidase Strip 100/Pk. Pack of 100
10726983 Microbact Reagent VP II 10 mL/Bottle 10mL
10331743 Microbact Oxidase Strip 50/Pk. 50 tests
12769030 Microbact Holding Tray 12A/12B 1 Tray Each
10036932 Microbact Reagent VP I 10 mL/Bottle 10mL
10371783 Microbact Reagent TDA 10 mL/Bottle 10mL
12749020 Microbact Reagent Set D 6 x 10 mL Bottles Each
10261063 Microbact Reagent Nitrate B 10 mL/Bottle 10mL
13296189 Microbact Holding Tray 24E/12E 1 Tray Each
13206189 Microbact Reagent Spot Indole DMACA 10 mL/Bottle 10mL
13245769 Microbact Mineral Oil 1 Bottle 50mL
10600975 Microbact Reagent Indole-Kovacs 10 mL/Bottle 10mL
10158442 Microbact Reagent Nitrate A 10 mL/Bottle Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 17108234 Supplier Thermo Scientific™ Supplier No. MB0266B
  • ONLINE EXCLUSIVE PRICE
    £168.14
    £91.15 / Pack of 100
    Save £76.99
    45% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.

Gram-negative bacteria are of significant concern in both clinical infections and food contamination. Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With these GNB reagents and accessories, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast treatment for the patient may follow.

  • Fast: More results available overnight
  • Simple: Simple test strip or microplate format
  • Easy to Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
  • Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
  • Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4

Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Identification Package (Windows™) (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The 6 reagents above are also available as Reagent Set D (MB1082A).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Description Microbact Oxidase Strip
Format Microplate, Strip
Packaging Type 1 Box
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.