missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Membrane Clostridium Perfringens Selective Supplement
Description
Oxoid Membrane Clostridium Perfringens Selective Supplement (m-CP) is a chromogenic medium for rapid and presumptive identification of Clostridium perfringens from water samples.
- Add to Membrane Clostridium Perfringens (m-CP) Agar Base, Part No. CM0992B
- Each vial of SR0188E supplements 500mL of medium
Specifications
Specifications
| Description | Membrane Clostridium Perfringens Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Perfringens |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?