missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Legionella MWY Selective Supplement

Product Code. 17150671
Change view
Click to view available options
Yield:
Each vial for 100mL medium
Each vial for 500mL medium
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Yield Unit Size
17150671 Each vial for 100mL medium Pack of 10
17140671 Each vial for 500mL medium Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17150671 Supplier Thermo Scientific™ Supplier No. SR0255E
  • ONLINE EXCLUSIVE PRICE
    £105.00
    £96.29 / Pack of 10
    Save £8.71
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.

Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.

Add to Legionella CYE Agar Base, Part No. CM0655B
TRUSTED_SUSTAINABILITY

Specifications

Description Legionella MWY Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 100mL medium
Species Legionella
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.