missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Legionella Agar Base

Product Code. 17170661
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17170661 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17170661 Supplier Thermo Scientific™ Supplier No. CM1203B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate Legionella from environmental samples and clinical specimens using this medium.

Isolate Legionella from environmental samples and specimens using Thermo Scientific™ Oxoid™ Legionella CYE Agar Base when used with Legionella Growth Supplement SR0251A. Legionella CYE Agar Base is a non-selective medium that exhibits optimal recovery of legionellaceae in short incubation periods from both environmental samples and specimens.

Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Transmission occurs through aerosolization or aspiration of contaminated water, not person to person.

Legionellosis refers to two distinct clinical syndromes, Legionnaires’ disease, which most often presents as severe pneumonia, and Pontiac fever, which is an acute, febrile, self-limiting, viral-like illness. Over forty species of Legionella have been characterized, however Legionella pneumophila remains the most prevalent etiological agent of Legionnaires’ disease.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: ISO Standards.
Description Legionella CYE Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 20.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.