missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Legionella BCYE Supplement

Product Code. 17180661
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Yield:
Each vial for 100mL medium
Each vial for 500mL medium
missing translation for 'unitSize'
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Yield unitSize
17180661 Each vial for 100mL medium Pack of 10
17190661 Each vial for 500mL medium Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 17180661 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' SR0251A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Legionella BCYE Supplement is used for the isolation of Legionella spp.

Oxoid Legionella BCYE Supplement is used for the isolation of Legionella spp.

Add to Legionella Agar Base, Part No. CM1203B
TRUSTED_SUSTAINABILITY

Specifications

Description Legionella BCYE Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 100mL medium
Species Legionella
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.