missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kanamycin Aesculin Azide Agar Base (Dehydrated)

Product Code. 10107662
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10107662 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10107662 Supplier Thermo Scientific™ Supplier No. CM0591B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect and isolate enterococci in foodstuffs with this selective medium.

Detect and isolate Enterococci in foodstuffs with Thermo Scientific™ Oxoid™ Kanamycin Aesculin Azide Agar Base (Dehydrated) when used with Kanamycin Supplement (SR0092). The medium contains the selective inhibitory components kanamycin sulphate and sodium azide. It also contains an indicator system to detect the growth of aesculin-hydrolysing streptococci. These organisms produce black zones around the colonies from the formation of black iron phenolic compounds derived from aesculin-hydrolysis products and ferrous iron.

Enterococci are part of the normal intestinal flora of human and animals. They have been long recognized as important human pathogens and are becoming increasingly so. Kanamycin Esculin Azide Agar is formulated as per Mossel et al 1, 2 to detect enterococci in foodstuffs. Mossel et al3 used it for the dip slide technique for bacteriological monitoring of foods.

  • Selective: due to the presence of kanamycin sulphate and sodium azide medium provides selectivity against non-target organisms.
  • Easy to read: aesculin-hydrolyzing streptococci form black zones around the colonies.
  • Can be used successfully for the isolation of glycopeptide resistant enterococci from foods4,5

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Kanamycin Aesculin Azide Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.7L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.