missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Iso-Sensitest Broth (Dehydrated)

Product Code. 10311053
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10311053

Brand: Thermo Scientific™ CM0473B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless antimicrobial susceptibility testing using this semi-defined medium specifically designed to give reproducible results.

Perform effortless testing using Thermo Scientific™ Oxoid™ Iso-Sensitest Broth (Dehydrated), a semi-defined medium specifically designed for antimicrobial susceptibility testing (AST). Iso-Sensitest Broth is carefully constructed to give reproducible results. It allows the growth of the majority of microorganisms without the need of further supplementation.

Iso-Sensitest Broth has stability in mineral content and the ability to produce optimum antimicrobial zones of inhibition. The amino-nitrogen base of acid-hydrolysed casein and special peptones has been supplemented with defined growth factors. Careful preparation of the nutrients ensures that trimethoprim and sulphonamide antagonists are at very low levels.

Iso-Sensitest Broth is produced in parallel with Thermo Scientific™ Oxoid™ Iso-Sensitest Agar CM0471. The broth has an identical nutrient formulation without the specifically purified agar. Where studies on antimicrobial susceptibilities are to be made in both broth and agar, this medium is found to be of particular value.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Iso-Sensitest Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 21.4L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.