missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Iron Sulphite Agar (Dehydrated)

Product Code. 10536893
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10536893 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10536893 Supplier Thermo Scientific™ Supplier No. CM0079B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect thermophilic anaerobic organisms causing sulphide spoilage in food with this medium.

Detect thermophilic anaerobic organisms causing sulphide spoilage in food with Thermo Scientific™ Oxoid™ Iron Sulphite Agar (Dehydrated). The medium is a modification of Cameron Sulphite Agar developed by the National Canners Association of America1. Iron Sulphite Agar has a reduced concentration of sodium sulphite to allow improved detection of some strains of Clostridium sporogenes.

Tryptone in the medium provides nitrogen and other nutrients necessary to support bacterial growth. The presence of sulphite reducing bacteria is indicated by the formation of black colonies. These colonies form when bacteria reduce sulphite to sulphide, which reacts with iron (III) citrate to yield a black precipitate.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Iron Sulphate Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 21.7L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.