missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO)

Product Code. 17851515
Change view
Click to view available options
Quantity:
500 g
5 kg
Yield:
7 L
70 L
Unit Size:
5kg
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Yield unitSize
17851515 500 g 7 L Each
18465620 5 kg 70 L 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17851515 Supplier Thermo Scientific™ Supplier No. CM1212B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate, enumerate and presumptively identify Listeria species and L. monocytogenes with this chromogenic medium.

Isolate, enumerate and presumptively identify Listeria species and L. monocytogenes from food and environmental samples with the Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) (Dehydrated culture medium and supplements) following broth enrichment. The medium identifies Listeria species based on their utilization of X-glucoside giving rise to blue-green colonies. Listeria monocytogenes are further differentiated by their ability to highly produce lecithinase, producing an opaque, white halo around the colony.

Isolate, enumerate and presumptively identify Listeria species and L. monocytogenes with Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Base.

  • Prepare Oxoid Brilliance Listeria Agar (ISO) Base by adding Thermo Scientific™ Brilliance Listeria Agar (ISO) Selective Supplement (SR0257) and Thermo Scientific™ Oxoid™ Brilliance Listeria Agar (ISO) Differential Supplement (SR0258) to Oxoid Brilliance Listeria  Agar (ISO).
  • Use as part of the Thermo Scientific™ Listeria Precis™ Detection and Enumeration method, or as part of the ISO 11290-1:2017 and ISO 11290-2:2017 reference methods for the detection and enumeration of Listeria species and L. monocytogenes.
  • Demonstrated to produce clearly differentiated colonies of Listeria within 22 hours when following the protocols for the Listeria Precis Methods or ISO 11290-1 and ISO 11290-2 methods.

Oxoid Brilliance Listeria Agar uses the chromogen X-glucoside for presumptive identification of Listeria spp. This chromogen is cleaved by ß-glucosidase which is common to all Listeria species. Other organisms that possess this enzyme, such as enterococci, are inhibited by the selective agents within the medium; lithium chloride, polymyxin B and nalidixic acid, whilst amphotericin inhibits the growth of any yeasts and molds present in the sample. Listeria monocytogenes and pathogenic Listeria ivanovii are then further differentiated by their ability to produce the phospholipase enzyme, lecithinase. This enzyme hydrolyses the lecithin in the medium, producing an opaque white halo around the colony.

The formulation and performance of Oxoid Brilliance Listeria Agar (ISO) conforms with ISO 11290-1:2017 and ISO 11290-2:2017 reference methods for the detection and enumeration of Listeria species and L. monocytogenes.

Oxoid BrillianceListeria Agar (ISO) Base has been validated as part of the Thermo Scientific™ Listeria Precis™ Methods for a broad range of samples according to ISO 16140-2 following NF validation rules when compared to the ISO 11290-1:2017 and ISO 11290 -2:2017.

TRUSTED_SUSTAINABILITY

Specifications

Description Thermo Scientific™ Oxoid™ Brilliance™ Listeria (ISO) Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield 7 L
Species Listeria
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.