missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Hemoglobin Soluble Powder

Product Code. 15660421
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
400g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15660421 1 Each 400g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15660421 Supplier Thermo Scientific™ Supplier No. LP0053C

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Oxoid Hemoglobin Powder Soluble is a specially prepared hemoglobin for use in culture media.

Oxoid Hemoglobin Powder Soluble is a specially prepared hemoglobin for use in culture media.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Shipped at room temperature. Store at 10° to 30°C. Keep tightly closed.
Description Dehydrated; Hemoglobin powder
Form Powder
Product Type Hemoglobin
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.