Learn More
Thermo Scientific™ Oxoid™ Helicobacter Pylori Selective Supplement (Dent)
Oxoid Helicobacter Pylori Selective Supplement (Dent) is for the isolaton of Helicobacter pylori from clinical specimens.
Brand: Thermo Scientific™ SR0147E
146.41 GBP valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Code : 65
Additional Details : Weight : 2.72160kg
Description
Oxoid Helicobacter Pylori Selective Supplement (Dent) is for the isolaton of Helicobacter pylori from clinical specimens.
- Used with Columbia Blood Agar Base, Part No. CM0331B
- Each vial supplements 500mL of medium
Specifications
Helicobacter pylori | |
Freeze-dried | |
10/Pk. |
Helicobacter Pylori Selective Supplement (Dent) | |
Selective Supplement | |
Each vial for 500mL medium |