missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Half Fraser Supplement

Product Code. 13235399
Change view
Click to view available options
Quantity:
10/Pk.
For 2.25L of base
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13235399 For 2.25L of base Pack of 10
11484209 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13235399 Supplier Thermo Scientific™ Supplier No. SR0166G
  • ONLINE EXCLUSIVE PRICE
    £310.07
    £258.26 / Pack of 10
    Save £51.81
    16% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.

Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.

  • Add to Fraser Broth Base, Part No. CM0895B
TRUSTED_SUSTAINABILITY

Specifications

Description Half Fraser Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity For 2.25L of base
Species Listeria
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.