missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Haemophilus Test Medium Base (Dehydrated)

Product Code. 11983902
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11983902

Brand: Thermo Scientific™ CM0898B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carry out susceptibility testing of Haemophilus influenzae with highly reproducible results with this medium.

Carry out susceptibility testing of Haemophilus influenzae with Thermo Scientific™ Oxoid™ Haemophilus Test Medium Base (Dehydrated). The medium is based on the formulation developed by Jorgensen et al1 which is now recommended by United States CLSI (M100Ed 29). The results achieved using Haemophilus Test Medium Base are highly reproducible3,4.

Haemophilus species are nutritionally fastidious in nature and require complex media for growth. Because of the antagonism between some essential nutrients and certain antimicrobial agents, the complex media formulations make routine susceptibility testing of Haemophilus species difficult. Regular Mueller Hinton Agar requires the addition of blood to supply essential nutrients making medium opaque, rendering it unsuitable for antimicrobial susceptibility testing.

Haemophilus Test Medium is based on the formulation developed by Jorgensen et al.2 which is now recommended by the United States CLSI. The transparency of the medium allows zones of inhibition to be read easily through the bottom of the Petri dish. Haemophilus Test Medium Base contains low levels of antimicrobial antagonists, which allows testing of trimethoprim/sulphamethoxazole to be carried out with greater confidence.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Haemophilus Test Medium Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 11.6L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.