Learn More
Thermo Scientific™ Oxoid™ GC Agar Base (Dehydrated)
Description
Selectively isolate Neisseria species with this medium containing meat and plant enzymatic digests.
Selectively isolate Neisseria species with Thermo Scientific™ Oxoid™ GC Agar Base (Dehydrated). GC Agar Base has been formulated to include Special Peptone LP0072 which is a mixture of meat and plant enzymatic digests. The presence of starch in the medium ensures that toxic metabolites produced by Neisseria are absorbed. Phosphate buffers are included to prevent changes in pH due to amine production that would affect the survival of the organism.
GC Agar Base is employed as a basal medium in the preparation of Chocolate Agar, Thayer-Martin Medium, Modified Thayer-Martin Medium, Martin-Lewis Agar, and Transgrow Agar.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | GC Agar Base |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 13.9L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.