Learn More
Thermo Scientific™ Oxoid™ Fraser Supplement
Product Code. 11434209
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 11434209
Brand: Thermo Scientific™ SR0156E
Edge
Oxoid Fraser Supplement is used for the isolation of Listeria species.
Oxoid Fraser Supplement is used for the isolation of Listeria species.
  • Add to Fraser Broth Base, Part No. CM0895B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Fraser Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium