Learn More
Thermo Scientific™ Oxoid™ Dextrose Bacteriological
Oxoid Dextrose Bacteriological is a sugar suitable for inclusion in bacteriological culture media.
Brand: Thermo Scientific™ LP0071B
33.31 GBP valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Code : 64
Additional Details : CAS Number : 5996-10-1 Weight : 0.53370kg
Description
Oxoid Dextrose Bacteriological is a sugar suitable for inclusion in bacteriological culture media.
Specifications
Dehydrated; Dextrose bacteriological | |
Powder | |
Media Supplement | |
500 g |