Learn More
Thermo Scientific™ Selenite Cystine Broth Base (Dehydrated)
Enrich and improve the isolation of salmonellae from feces and food samples with this medium.
£77.20
Specifications
Content And Storage | 10°C to 30°C |
---|---|
Description | Selenite Cystine Broth Base (Dehydrated) |
Form | Powder |
Product Type | Dehydrated Culture Media |
Description
Enrich and improve the isolation of salmonellae from feces and food samples with Thermo Scientific™ Oxoid™ Selenite Cystine Broth Base. This modified medium is based upon the formulation described by Leifson1 with added cystine.2
Selenite Cystine Broth Base is an enrichment medium for the isolation of salmonellae from feces and food products.
- Cystine reduces the toxicity of selenite and provides extra organic sulphur, which spares critical sulphur components of the bacteria
- Formulation recommended by the AOAC3 for detection of Salmonella in foodstuffs
- Included in the standard methods media of the American Public Health Association4, 5
- Complies with the requirements of the United States Pharmacopoeia6
Selenite Cystine Broth Base is a modification of the formulation described by Leifson1 with cysteine and is used as an enrichment culture for salmonellae from feces, foodstuffs, and other materials.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
10°C to 30°C | |
Selenite Cystine Broth Base (Dehydrated) | |
Powder | |
Dehydrated Culture Media |