Learn More
Thermo Scientific™ Oxoid™ Defibrinated Sheep Blood
Description
Defibrinated Sheep Blood is a non-selective, general-purpose enrichment supplement that may be used in the preparation of many types of medium for the isolation and culture of pathogenic and non-pathogenic microorganisms from clinical samples (e.g. cerebral spinal fluid, pus and wound swabs, sputum, eye swabs and ear, nose and throat swabs, blood and fecal samples).
Key Benefits and Product Features
- General-purpose enrichment: Suitable for preparing blood agar and other enriched culture media for routine microbiology.
- Supports microbial growth: Enhances the isolation and cultivation of pathogenic and non-pathogenic microorganisms from clinical specimens.
- Hemolysis detection: Enables observation of hemolytic reactions, colony morphology, and hemolysis patterns for microbial identification.
- Compatible with blood agar media: Ideal for use with Columbia Blood Agar (CBA), CNA, and other blood agar formulations.
- Defibrinated: Mechanically processed to remove fibrin, preventing clot formation while maintaining blood quality.
- Aseptically collected: Manufactured under controlled conditions to ensure sterility and consistent performance.
- Clinical applications: Suitable for culturing microorganisms from cerebrospinal fluid, blood, sputum, pus, wound swabs, eye swabs, ear, nose and throat swabs, and fecal specimens.
- Storage: Store in the original packaging between 2°C and 8°C.
Specifications
Specifications
| Content And Storage | Store product in its original packaging between 2°C and 8°C |
| Species | Sheep |
| Form | Liquid |
| Quantity | 1 x 100 mL |
| Processing | Defibrinated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.