Learn More
Thermo Scientific™ Czapek Dox Liquid Medium (Modified) (Dehydrated)
Description
Cultivate those fungi and bacteria which can utilize sodium nitrate as the sole source of nitrogen using Thermo Scientific™ Oxoid™ Czapek Dox Liquid Medium (Modified) (Dehydrated).
Use Czapek Dox Liquid Medium (Modified) for cultivation of those fungi and bacteria which can utilize sodium nitrate as the sole source of nitrogen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Czapek Dox Liquid Medium |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 15.0L medium |
For Laboratory Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.