missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cold Filterable Tryptone Soya Broth (Dehydrated)

Product Code. 10462263
Change view
Click to view available options
Quantity:
25 kg
5 kg
500 g
Each
Unit Size:
2.5kg
25kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10462263 500 g 500g
10631105 5 kg 5kg
12606918 25 kg 25kg
12570836 Each 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10462263 Supplier Thermo Scientific™ Supplier No. CM1065B

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Carry out microbiological Media Fill Trials (MFT) in the pharmaceutical industry with this gamma-irradiated medium.

Carry out microbiological Media Fill Trials (MFT) in the pharmaceutical industry with gamma-irradiated Thermo Scientific™ Oxoid™ Cold Filterable Tryptone Soya Broth (Dehydrated). The highly nutritious general purpose medium conforms to the formulation stated for MFT in the European1, British2, US3 and Japanese4 Pharmacopoeias.

Cold Filterable Tryptone Soya Broth can be substituted for the powdered components that go into making aqueous drug products to provide an index of the level of microbial contamination occurring in aseptic manufacture.

‘Sterile for use’ liquid drugs often contain heat-sensitive components which means that terminal sterilisation by autoclaving is not an option. Sterilisation by filtering (for soluble liquids) followed by filling under aseptic conditions is the method for preparation of these types of drug. The purpose of Media Fill Trials is to provide a measure of the likelihood of microbiological contamination arising in a particular aseptic process.

Packs of Cold Filterable Tryptone Soya Broth have been given a sterilizing dose of gamma-irradiation (minimum 25KGy) validated as a lethal dose for all yeasts, molds, bacterial spores and mycoplasma.

  • Gamma-irradiated to ensure that the medium is free from all yeasts, moulds, and bacteria, including bacterial spores and mycoplasmas, prior to use.
  • Screened and selected to ensure a highly filterable solution that has a known Vcap value. This allows the maximum filterable volume to be calculated before starting a MFT.

The purpose of MFT is to provide a measure of the likelihood of microbiological contamination occurring in a particular aseptic process. Cold Filterable TSB can be used as a substitute for filter-sterilized drug products and is processed in a manner identical to that in which the product would be processed i.e. filtering, filling and closing. The medium is then incubated and the number of contaminated units is scored versus those that are un-contaminated. A decision following pre-determined guidelines can be made based on the proportion of contaminated units and the identity of the microorganisms recovered5.

Cold Filterable Tryptone Soya Broth may also be used to test aseptic production processes for solid presentations by adding the medium as a sterile liquid downstream of processing a placebo5.

For laboratory use, standard Thermo Scientific™ Oxoid™ Tryptone Soya Broth (CM0129) may be more appropriate.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Cold Filterable Tryptone Soya Broth
Form Powder
Media Type Tryptone Soya Broth
Product Type Nutrient Broth
Quantity 500 g
Yield For 16.7L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.