missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Ceftolozane/Tazobactam Antimicrobial Susceptibility discs

Product Code. 17979012
This item is not returnable. View return policy
Product Code. 17979012 Supplier Thermo Scientific™ Supplier No. CT1970B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform manual susceptibility testing of multi-drug and extensively drug resistant bacteria with proven quality1, easy-to-use AST discs which meet EUCAST and CLSI breakpoints

Manually determine the antibiotic susceptibility of microorganisms using disc diffusion methods with consistently reliable, industry leading quality1 Thermo Scientific™ Oxoid™ Ceftolozane/Tazobactam Antimicrobial Susceptibility discs. The drug ceftolozane-tazobactam has been designed to address both Gram negative and Gram positive (in cIAI) that are multi-drug (MDR) or extensively drug (XDR) resistant. Ceftolozane-tazobactam is aimed at the treatment of serious infections, such as complicated UTIs, complicated IAIs and bacterial pneumonia.

Perform susceptibility testing of multi-drug and extensively drug resistant organisms using Oxoid™ Ceftolozane/Tazobactam Antimicrobial Susceptibility discs which contain the cephalosporin, ceftolozane, and the novel beta-lactamase inhibitor, tazobactam.

  • Meets EUCAST and CLSI recommended breakpoints
  • Easy-to-use: shatterproof spring-loaded cartridges, easy-to-read labelling, and end-of-cartridge indicators.
  • Long-term stability ensured: each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%.
  • Accurate and reproducible: manufactured under stringent quality standards including BS EN ISO 9001:2008, ISO 13485:2016 and FDA 21 CFR part 820, with comprehensive quality control testing.
  • Trusted supplier: over 35 years of experience in disc development and manufacture

disc diffusion not only categorizes resistant, intermediate, and sensitive organisms through qualitative results but also provides a visual indication of the following:

  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics

Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Ceftolozane, Tazobactam
Concentration 30/10 μg
Description Oxoid™ Ceftolozane/Tazobactam Antimicrobial Susceptibility Discs
Antibiotic Code C/T40
Product Type Antimicrobial Susceptibility Discs
Target Organism Class Multidrug-resistant, Gram-negative Organisms, Multidrug-resistant, Gram-positive Organisms
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge

Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.