Learn More
Thermo Scientific™ Oxoid™ Cefixime Supplement
Description
Oxoid Cefixime Supplement is used for the isolation of Escherichia coli O157.
- Add to to Cefixime Rhamnose Sorbitol MacConkey Agar Base, Part No. CM1005B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Cefixime Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |