missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ CCDA Selective Supplement
Oxoid Brain Heart Infusion Broth is highly nutritious medium for the growth of fastidious organisms, recommended for blood culture work; also see Heart Infusion Broth.
£243.00 - £365.00
Specifications
Description | CCDA Selective Supplement |
---|---|
Form | Freeze-dried |
Product Type | Selective Supplement |
Product Code | Brand | Quantity | Yield | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Yield | Price | Quantity & Availability | ||||
11424209
|
Thermo Scientific™
SR0155E |
10/Pk. | Each vial for 500mL medium |
£243.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
13205399
|
Thermo Scientific™
SR0155H |
10 Vials | For 2L medium |
£365.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
Description
Oxoid CCDA Selective Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Arcobacter.
- For the isolation of Arcobacter spp., add to Arcobacter Broth Base, Part No. CM0965B
- For the isolation of Campylobacter spp., especially C. upsaliensis, add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B
Specifications
CCDA Selective Supplement | |
Freeze-dried | |
Selective Supplement |