missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ CCDA Selective Supplement

Product Code. 11424209
Click to view available options
Quantity:
10 Vials
10/Pk.
Yield:
Each vial for 500mL medium
For 2L medium
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11424209

Brand: Thermo Scientific™ SR0155E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Brain Heart Infusion Broth is highly nutritious medium for the growth of fastidious organisms, recommended for blood culture work; also see Heart Infusion Broth.

Oxoid CCDA Selective Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Arcobacter.

  • For the isolation of Arcobacter spp., add to Arcobacter Broth Base, Part No. CM0965B
  • For the isolation of Campylobacter spp., especially C. upsaliensis, add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B
TRUSTED_SUSTAINABILITY

Specifications

Description CCDA Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.