missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ CAT Supplement

Product Code. 12742020
Click to view available options
Quantity:
10/Pk.
For 2L medium
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 12742020

Brand: Thermo Scientific™ SR0174H

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Oxoid CAT Supplement isolates Campylobacter spp.

Oxoid CAT Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Acrobacter.

  • Add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B for the isolation of Campylobacter spp.
  • Add to Arcobacter Broth Base, Part No. CM0965B for the isolation of Arcobacter spp.
TRUSTED_SUSTAINABILITY

Specifications

Description Cat Sel Suppl For 2l 10/Pk.
Form Freeze-dried
Product Type Microbiology Media
Quantity For 2L medium
Species Campylobacter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.