missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ CAT Supplement
Click to view available options
Quantity:
10/Pk.
For 2L medium
Unit Size:
Each
Pack of 10
Description
Oxoid CAT Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Acrobacter.
- Add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B for the isolation of Campylobacter spp.
- Add to Arcobacter Broth Base, Part No. CM0965B for the isolation of Arcobacter spp.
Specifications
Specifications
| Description | CAT Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Campylobacter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction