Learn More
Thermo Scientific™ Oxoid™ Casein Hydrolysate
Oxoid Casein Hydrolysate (Acid), a purified hydrolysate, provides nitrogenous material for culture media or can be used when amino acid mixtures are specified.
Brand: Thermo Scientific™ LP0041B
165.71 GBP valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Code : 04
Additional Details : Weight : 0.59300kg
Description
Oxoid Casein Hydrolysate (Acid) is a purified hydrolysate which provides the necessary nitrogenous material for culture media or can be used when amino acid mixtures are specified.
Specifications
Dehydrated; Casein hydrolysate (Acid) | |
Powder | |
Hydrolysate | |
500 g |