missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Campylobacter Karmali Agar Base (Dehydrated)

Product Code. 10612065
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10612065 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10612065 Supplier Thermo Scientific™ Supplier No. CM0935B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate Campylobacter jejuni and Campylobacter coli from clinical specimens with this selective medium.

Isolate Campylobacter jejuni and Campylobacter coli using Thermo Scientific™ Oxoid™ Campylobacter Karmali Agar Base (Dehydrated) when used with Modified Karmali Selective Supplement (SR0205) or Campylobacter Selective Supplement (Karmali) (SR0167). Campylobacter Karmali Agar Base is based on the formulation described by Karmali et al.1 and is recommended for the isolation of Campylobacter jejuni and Campylobacter coli from clinical specimens.

Campylobacter are Gram-negative, spirally shaped microaerophilic organisms which may be present in raw milk, untreated water, improperly handled food and undercooked meats, poultry and shellfish. Human consumption of these organisms can result in a range of clinical illnesses from transient asymptomatic colonization to severe dysentery. The symptoms of Campylobacter enteritis include diarrhoea, stomach pain, nausea, fever, headache and muscle pain.

Use Campylobacter Karmali Agar Base for isolation of Campylobacter species.

  • Selective: due to the presence of the selective agents like vancomycin and amphotericin B the medium is selective for Campylobacter
  • Easy-to-read: Campylobacter jejuni strains produce grey, moist, flat spreading colonies after 42-hour incubation at 42°C.

Campylobacter Karmali Agar Base is a modification of the original Campylobacter Blood Free medium. Campylobacter Blood Free medium contains sodium pyruvate in the agar base. Campylobacter Medium (Karmali) incorporates this ingredient into the selective supplement. The original medium also contains sodium desoxycholate for the inhibition of Gram-positive organisms, whereas with Campylobacter Medium (Karmali) suppression of Gram-positives is achieved by the inclusion of vancomycin.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Campylobacter Agar Base (Karmali)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.6L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.