missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Brucella Selective Supplement
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
Oxoid Brucella Selective Supplement is used for the isolation of Brucella spp.
- Add to Blood Agar Base No. 2, Part No. CM0271B; Columbia Blood Agar Base, Part No. CM0331B; or Brucella Medium Base, Part No. CM0169B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Brucella Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Brucella species |
Safety and Handling
Product Identifier
- Brucella Selective Supplemen
- Danger
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Skin sensitiser Category 1
- H300-Fatal if swallowed.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H360-May damage fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P363-Wash contaminated clothing before reuse.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Cycloheximide, Vancomycin hydrochloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction