missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brucella Medium Base (Dehydrated)

Product Code. 12980219
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 12980219

Brand: Thermo Scientific™ CM0691B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prepare the serum-dextrose-antibiotic medium described by Jones and Brinley Morgan1 for the cultivation and isolation of Brucella with this medium.

Prepare the serum-dextrose-antibiotic medium described by Jones and Brinley Morgan1 for the cultivation and isolation of Brucella with Thermo Scientific™ Oxoid™ Brucella Medium Base (Dehydrated) when used with Brucella Selective Supplements SR0083 or SR0209. Without the selective supplements, the medium can be used in conjunction with Cruickshank dyestrip method for differentiation between strains.

Use Brucella Medium Base for the preparation of serum-dextrose-antibiotic medium for the cultivation and isolation of Brucella using Brucella Selective Supplement SR0083 or SR0209. Without the supplements, it may be used in conjunction with the Cruickshank dyestrip method for differentiation between strains.

Brucella medium with supplements is more effective as a selective medium over the media described by Kuzdas and Morse2 and Renoux3 as it supports the growth of fastidious types.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Brucella Agar (USA)
Form Powder
Product Type Agar
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.