Learn More
Thermo Scientific™ Brilliance™ Candida Agar Base (Dehydrated)
Description
Rapidly isolate and identify clinically important Candida species with this medium.
Rapidly isolate and identify clinically important Candida species with Thermo Scientific™ Brilliance™ Candida Agar Base (Dehydrated). This chromogenic formulation produces four different diagnostic colors for the presumptive identification of clinically significant Candida species and provides faster, easier-to-read results when compared to traditional media.
Serious infections due to Candida species are becoming increasingly prevalent. This poses particular problems because of the increasing incidence of non-albicans spp. and the emergence of non-albicans isolates resistant to both amphotericin B and the newer azoles.1
Isolate, differentiate and identify Candida albicans and Candida tropicalis from other Candida spp. within 48 hours with this selective differential chromogenic medium.
- Rapid: allows the identification of Candida albicans and Candida tropicalis from other species of Candida within 48 hours.
- Easy-to-read:Candidaalbicans (green colonies) is easy to isolate from Candida tropicalis (dark blue colonies)
Brilliance Candida Agar incorporates two chromogens that indicate the presence of the target enzymes:
- X-NAG (5-bromo-4-chloro-3-indolyl N acetyl ß-D-glucosaminide) detects the activity of hexosaminidase.
- BCIP (5-bromo-6-chloro-3-indolyl phosphate p-toluidine salt) detects alkaline phosphatase activity
The typical enzyme patterns of Candida spp. as below:
| Chromogen: Enzyme: | X-NAG Hexosaminidase | BCIP Alkaline phosphatase | Typical colony appearance |
| C. tropicalis | + | Dark blue | |
| C. albicans C. dubliniensis | + | Green ‡ | |
| C. krusei | + | Dry, irregular pink-brown | |
| C. glabrata C. kefyr C. parapsilosis C. lusitaniae | Variable | Beige/yellow/ brown † |
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Brilliance Candida Agar Base |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 16.0L medium |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.