missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Bordetella Selective Supplement
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
Oxoid Bordetella Selective Supplement (Cephalexin, 20mg/vial) is used for the selective isolation of Bordetella pertussis and B. parapertussis.
Add to to Charcoal Agar Base, Part No. CM0119B. Each vial supplements 500mL of medium.
Specifications
Specifications
| Description | Bordetella Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Bordetella species |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction