missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Bolton Broth Selective Supplement

Product Code. 11474219
Click to view available options
Quantity:
10/Pk.
Unit Size:
175g
This item is not returnable. View return policy

Product Code. 11474219

Brand: Thermo Scientific™ SR0183E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Bolton Broth Selective Supplement is used for the selective pre-enrichment of Campylobacter spp. in food samples.

Oxoid Bolton Broth Selective Supplement is used for the selective pre-enrichment of Campylobacter spp. in food samples.

Add to to Bolton Broth, Part No. CM0983B. Each vial supplements 500mL of medium.
TRUSTED_SUSTAINABILITY

Specifications

Description Bolton Broth Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Bolton
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.