missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BiGGY Agar (Nickerson Medium) (Dehydrated)

Product Code. 10780894
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10780894 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10780894 Supplier Thermo Scientific™ Supplier No. CM0589B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and presumptively identify Candida species with this selective medium.

Isolate and presumptively identify Candida species with Thermo Scientific™ Oxoid™ BiGGY Agar (Nickerson Medium) (Dehydrated). BiGGY, Bismuth Sulphite Glucose Glycine Yeast Agar, is based on the formulation developed by Nickerson1 and is a highly selective medium inhibiting most of the competing flora.

Serious infections due to Candida species are becoming increasingly prevalent. This poses particular problems because of the increasing incidence of non-albicans spp. and the emergence of non-albicans isolates resistant to both amphotericin B and the newer azoles2.

Use BiGGY Agar for isolation and identification of the Candida species.

  • Selective: the presence of bismuth sulphite complex in the medium confers a high degree of selectivity to the medium, and most strains of bacteria are inhibited on BIGGY Agar.
  • Can be used for the isolation and presumptive identification of Candida albicans and Candida tropicalis from sputum3,4 and vaginal smears5
  • Recommended medium for the quality assessment of pharmaceutical and cosmetic products6

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description BiGGY Agar (Nickerson Medium)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.9L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.