Learn More
Thermo Scientific™ Azide Dextrose Broth (Dehydrated)
Description
Detect enterococci in water and sewage samples with this selective medium.
Detect enterococci in water and sewage samples with Thermo Scientific™ Oxoid™ Azide Dextrose Broth (Dehydrated). A blend of peptone and glucose in the medium renders Azide Dextrose Broth highly nutritious, and sodium chloride maintains the osmotic equilibrium. Sodium azide acts as an inhibitor of Gram-negative organisms1,2,3 and the concentration of sodium azide used in the medium provides optimum protection for the enterococci while largely suppressing the Gram-negative flora. The phosphate buffer system controls pH.
Enterococci are part of the normal intestinal flora of human and animals. They have been long recognized as important human pathogens and are becoming increasingly so. The presence of enterococci serves as an indicator of fecal contamination. Enterococci are better indicators than Escherichia coli of sewage pollution in chlorinated waters because they have a greater resistance to chlorine.
Azide Dextrose broth is recommended for the detection of enterococci in water and sewage4.
- Selective: sodium azinde in the medium acts as an inhibitor of Gram-negative organisms while providing optimum protection to enterococci.
- Recommended for the quantitative determination of enterococci in water, sewage, foods and other materials suspected of contamination with sewage1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Azide Dextrose Broth |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 14.0L medium |
Safety and Handling
- AZIDE DEXTROSE BROTH Rothe
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Sodium azide
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.