Learn More
Thermo Scientific™ Remel™ Antimicrobial Susceptibility disc Dispenser, 150 mm, 12-place
Product Code. 11943462
Change view
Click to view available options
Quantity:
1 Dispenser/Pk.
Unit Size:
Each
Product Code. Quantity unitSize
11943462 1 Dispenser/Pk. Each
1 options
Product Code. 11943462 Supplier Thermo Scientific™ Supplier No. ST1215
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods.
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Remel™ Antimicrobial Susceptibility disc Dispenser.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance For use only with Oxoid Antimicrobial Susceptibility Discs
Description Remel Antimicrobial Susceptibility Disc Dispenser
Product Type Antimicrobial Susceptibility Disc Dispenser
No. of Places 12-place
Quantity 1 Dispenser/Pk.