missing translation for 'onlineSavingsMsg'
Learn More

Oxoadipic acid, MedChemExpress

Product Code. 16424768
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16424768

Brand: Medchem Express HY113227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxoadipic acid is a key metabolite of the essential amino acids tryptophan and lysine.

Oxoadipic acid is a key metabolite of the essential amino acids tryptophan and lysine.

Specifications

Chemical Name or Material Oxoadipic acid
Physical Form Solid
CAS 3184-35-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C6H8O5
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information H2O : 250 mg/mL (1561.33 mM; Need ultrasonic)
SMILES O=C(O)C(CCCC(O)=O)=O
Molecular Weight (g/mol) 160.12
Formula Weight 160.12
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.