missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Oxivir Excel Disinfectant Wipes

Product Code. 17261614
Click to view available options
Dimensions (L x W):
15 x 17 cm
26 x 20 cm
27 x 20 cm
Format:
Canister, PT02
Flow Pack
Packaging Quantity:
100 Wipes/Pk., 24 Pk.
160 Wipes/Pk., 6 Pk.
30 Wipes/Pk., 12 Pk
Unit Size:
Pack of 2400
Pack of 360
Pack of 960
This item is not returnable. View return policy

Product Code. 17261614

Brand: Diversey™ 100984250

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pre-moistened cleaner disinfectant wipes for non-invasive medical devices and nonporous surfaces

  • One-step cleaner disinfectant with broad spectrum efficacy and short contact times
  • Oxivir Excel Wipe can be used on non-invasive medical devices and nonporous surfaces
  • Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 seconds
  • Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
  • No hazard classification
  • Powered by AHP Technology (Accelerated Hydrogen Peroxide)
  • AHP formulation is gentle on surfaces extending equipment life
  • Excellent user safety profile - no PPE required
  • Does not contain respiratory irritants
  • Suitable for use in high risk areas including ICU, OR, NICU and patient rooms
  • Fragrance free
  • No rinsing required
  • Safer for the environment and user, since the AHP actives break down into water and oxygen. Free from Volatile Organic Compounds (VOCs)
  • Medical Device Class IIa

Specifications

Product Type All Purpose Wipes
Length (English) 10 in.
Length (Metric) 26 cm
Width (English) 8 in.
Width (Metric) 20 cm
Material Polypropylene
Description For DK, FI, GB, IE, and SE only
Dimensions (L x W) 26 x 20 cm
Format Flow Pack
For Use With (Application) All purpose virucidal wipes
Packaging Quantity 30 Wipes/Pk., 12 Pk
Sterility Non-sterile
Certifications/Compliance (Europe) Efficacy proven by compliance with EN13727, EN13624, EN14476 and EN16615
Show More Show Less

The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.