missing translation for 'onlineSavingsMsg'
Learn More

Oxindole, MedChemExpress

Product Code. 16479406
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479406 Supplier Medchem Express Supplier No. HYY0061
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxindole (Indolin-2-one) is an aromatic heterocyclic building block. 2-indolinone derivatives have become lead compounds in the research of kinase inhibitors.

Oxindole (Indolin-2-one) is an aromatic heterocyclic building block. 2-indolinone derivatives have become lead compounds in the research of kinase inhibitors.

Specifications

Chemical Name or Material Oxindole
CAS 59-48-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C8H7NO
Quantity 100 mg
Synonym Indolin-2-one
Solubility Information DMSO : 100 mg/mL (751.03 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=CC=C2)C1
Molecular Weight (g/mol) 133.15
Formula Weight 133.15
Percent Purity 98.82%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.