missing translation for 'onlineSavingsMsg'
Learn More

Oxindole, 97+%

Product Code. 11463038
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463038 5 g 5g
11473038 25 g 25g
11483038 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463038 Supplier Thermo Scientific Alfa Aesar Supplier No. A18764.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-48-3 | C8H7NO | 133.15 g/mol

Oxindole is considered as indole analogue, which shows pharmacological activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxindole is considered as indole analogue, which shows pharmacological activity.

Solubility
Soluble in methanol, ethanol, dimethyl sulfoxide and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-48-3
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005711
InChI Key JYGFTBXVXVMTGB-UHFFFAOYSA-N
Synonym oxindole, indolin-2-one, 2-oxindole, 2-indolinone, 1,3-dihydro-2h-indol-2-one, oxindol, 2-oxoindoline, 2-oxindoline, indol-2 3h-one, 2h-indol-2-one, 1,3-dihydro
PubChem CID 321710
ChEBI CHEBI:31697
IUPAC Name 1,3-dihydroindol-2-one
SMILES C1C2=CC=CC=C2NC1=O

Specifications

Melting Point 124°C to 128°C
Boiling Point 194°C to 196°C (17 mmHg)
Quantity 5 g
Beilstein 114692
Solubility Information Soluble in methanol,ethanol,dimethyl sulfoxide and ether. Insoluble in water.
Formula Weight 133.15
Percent Purity ≥97%
Chemical Name or Material Oxindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.