missing translation for 'onlineSavingsMsg'
Learn More

Oxindole-3-acetic Acid, TRC

Product Code. 30414760
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414760 100 mg
30414759 250 mg
30413731 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414760 Supplier Toronto Research Chemicals Supplier No. TRCO850140100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2971-31-5
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.18
Synonym 2-Oxo-3-Indolineacetic Acid, (±)-2,3-Dihydro-2-oxoindole-3-acetic Acid, (±)-2-Oxindoline-3-acetic Acid, (±)-2-Oxo-3-indolineacetic Acid, 2-(2-Oxoindolin-3-yl)acetic Acid
IUPAC Name 2-(2-oxo-1,3-dihydroindol-3-yl)acetic acid
SMILES O=C1NC2=CC=CC=C2C1CC(O)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H9NO3/c12-9(13)5-7-6-3-1-2-4-8(6)11-10(7)14/h1-4,7H,5H2,(H,11,14)(H,12,13)
Formula Weight 191.0582
Chemical Name or Material Oxindole-3-acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.