missing translation for 'onlineSavingsMsg'
Learn More

Oxindole-3-acetic Acid-d4, TRC

Product Code. 30417039
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417039 10 mg 10mg
30421910 1 mg 1mg
30414999 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30417039 Supplier Toronto Research Chemicals Supplier No. TRCO85014210MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxindole-3-acetic Acid-d4
Molecular Formula C10 D4 H5 N O3
Quantity 10 mg
Synonym 2-Oxo-3-Indolineacetic-d4 Acid, (±)-2,3-Dihydro-2-oxoindole-3-acetic-d4 Acid, (±)-2-Oxindoline-3-acetic-d4 Acid, (±)-2-Oxo-3-indolineacetic-d4 Acid, 2-(2-Oxoindolin-3-yl)acetic-d4 Acid
InChI Formula InChI=1S/C10H9NO3/c12-9(13)5-7-6-3-1-2-4-8(6)11-10(7)14/h1-4,7H,5H2,(H,11,14)(H,12,13)/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c2C(CC(=O)O)C(=O)Nc2c1[2H]
IUPAC Name 2-(4,5,6,7-tetradeuterio-2-oxo-1,3-dihydroindol-3-yl)acetic acid
Molecular Weight (g/mol) 195.208
Formula Weight 195.083
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.