missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxilofrine hydrochloride, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Oxilofrine hydrochloride
Specifications
Specifications
| Chemical Name or Material | Oxilofrine Hydrochloride |
| CAS | 942-51-8 |
| Analyte or Component Names | Oxilofrine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H15 N O2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Oxilofrine Hydrochloride, Benzenemethanol, 4-hydroxy-alpha-[1-(methylamino)ethyl]-, hydrochloride, (R*,S*)- (9CI), 4-Hydroxyephedrinehydrochloride, Suprifen hydrochloride, p-Hydroxyephedrine hydrochloride |
| InChI Formula | InChI=1S/C10H15NO2.ClH/c1-7(11-2)10(13)8-3-5-9(12)6-4-8;/h3-7,10-13H,1-2H3;1H/t7-,10-;/m0./s1 |
| SMILES | Cl.CN[C@@H](C)[C@H](O)c1ccc(O)cc1 |
| IUPAC Name | 4-[(1R,2S)-1-hydroxy-2-(methylamino)propyl]phenol;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction