missing translation for 'onlineSavingsMsg'
Learn More

Oxidopamine hydrobromide, MedChemExpress

Product Code. 16458596
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458596 100 mg 100mg
16431456 200 mg 200mg
16414547 50 mg 50mg
16402256 500 mg 500mg
16482309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16458596 Supplier Medchem Express Supplier No. HYB1081A
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Oxidopamine (6-OHDA) hydrobromide is an antagonist of the neurotransmitter dopamine.

Oxidopamine hydrobromide is a widely used neurotoxin and selectively destroys dopaminergic neurons. Oxidopamine hydrobromide promotes COX-2 activation, leading to PGE2 synthesis and pro-inflammatory cytokine IL-1β secretion. Oxidopamine hydrobromide can be used for the research of Parkinson’s disease (PD), attention-deficit hyperactivity disorder (ADHD), and Lesch-Nyhan syndrome.

Specifications

Chemical Name or Material Oxidopamine hydrobromide
CAS 636-00-0
Color Gray
Purity Grade Notes Research
Molecular Formula C8H12BrNO3
Quantity 100 mg
Synonym 6-Hydroxydopamine hydrobromide 6-OHDA hydrobromide
Solubility Information DMSO : 50 mg/mL (199.93 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 20 mg/mL (79.97 mM; Need ultrasonic)
SMILES OC1=CC(CCN)=C(O)C=C1O.[H]Br
Molecular Weight (g/mol) 250.09
Formula Weight 250.09
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.