missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxidized PTP Active Site Antibody, R&D Systems™

Description
Oxidized PTP Active Site Monoclonal specifically detects Oxidized PTP Active Site in Multi-Species samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Oxidized PTP Active Site |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 335636 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Host Species | Mouse |
| Immunogen | KLH-coupled perfomic acid oxidized synthetic peptide VHCSAG |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?