missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OXGR1 Recombinant Rabbit Monoclonal Antibody (JE64-04)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544734
This item is not returnable.
View return policy
Description
OXGR1 Recombinant Monoclonal Antibody for Western Blot
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
Specifications
| OXGR1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6IYF8, Q6Y1R5, Q96P68 | |
| OXGR1 | |
| Synthetic peptide within human OXGR1 aa 171-220/337. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| JE64-04 | |
| Unconjugated | |
| OXGR1 | |
| 2-oxoglutarate receptor 1; aKGR; Alpha-ketoglutarate receptor 1; Cysltr3; G protein-coupled receptor 80; G protein-coupled receptor 99; Gm299; Gpr80; Gpr99; G-protein coupled receptor 80; G-protein coupled receptor 99; MGC119206; MGC119207; MGC119208; Oxgr1; oxoglutarate (alpha-ketoglutarate) receptor 1; oxoglutarate receptor 1; P2RY15; P2Y purinoceptor 15; P2Y15; P2Y-like GPCR; P2Y-like nucleotide receptor; seven transmembrane helix receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 239283, 27199, 290493 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction