missing translation for 'onlineSavingsMsg'
Learn More

OXGR1/GPR80/P2Y15 Antibody - BSA Free, Novus Biologicals™

Product Code. 18319964 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319964 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319964 Supplier Novus Biologicals Supplier No. NLS6747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OXGR1/GPR80/P2Y15 Polyclonal specifically detects OXGR1/GPR80/P2Y15 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXGR1/GPR80/P2Y15
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 19-38 ug/ml
Formulation PBS with 0.1% Sodium Azide
Gene Alias Alpha-ketoglutarate receptor 1, G protein-coupled receptor 80, G protein-coupled receptor 99, GPR802-oxoglutarate receptor 1, GPR99, G-protein coupled receptor 80, G-protein coupled receptor 99, MGC119206, MGC119207, MGC119208, oxoglutarate (alpha-ketoglutarate) receptor 1, P2RY15P2Y-like GPCR, P2Y purinoceptor 15, P2Y15P2Y-like nucleotide receptor, seven transmembrane helix receptor
Gene Symbols OXGR1
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-terminal extracellular domain of human OXGR1/GPR80/P2Y15.
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 27199
Test Specificity G Protein-Coupled Receptor GPR80/GPR99/P2Y15
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.