missing translation for 'onlineSavingsMsg'
Learn More

Oxfendazole, MedChemExpress

Product Code. 16488216
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488216 100 mg 100mg
16482086 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488216 Supplier Medchem Express Supplier No. HYB0291
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxfendazole is the sulfoxide form of fenbendazole which is a broad spectrum benzimidazole anthelmintic.

Oxfendazole is the sulfoxide form of fenbendazole which is a broad spectrum benzimidazole anthelmintic.

Specifications

Chemical Name or Material Oxfendazole
Physical Form Solid
CAS 53716-50-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H13N3O3S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 31.25 mg/mL (99.10 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 0.67 mg/mL (2.12 mM; Need ultrasonic)
SMILES O=C(OC)NC1=NC2=CC=C(S(C3=CC=CC=C3)=O)C=C2N1
Molecular Weight (g/mol) 315.35
Formula Weight 315.35
Percent Purity 99.26%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.